Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 18504 |
Gene Name: | paired box 2 |
Gene Aliases: |
Opdc, Pax-2 |
Location: |
Chr.19:44745695-44837871 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pax2 | NM_011037.4 | NP_035167.4 | ||
BC148232.1 | ||||
BC150484.1 |
Set Membership: |
Intragenic Intronic Non-exonic |