Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 9990 |
Gene Name: | solute carrier family 12 member 6 |
Gene Aliases: |
ACCPN, KCC3, KCC3A, KCC3B |
Location: |
Chr.15:34229996-34338064 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC12A6 | NM_001042494.1 | 21 | 3097 | NP_001035959.1 |
NM_001042495.1 | 21 | 2772 | NP_001035960.1 | |
NM_001042496.1 | 21 | 3163 | NP_001035961.1 | |
NM_001042497.1 | 19 | 2866 | NP_001035962.1 | |
NM_005135.2 | 20 | 2736 | NP_005126.1 | |
NM_133647.1 | 20 | 2911 | NP_598408.1 | |
XM_006720793.3 | 19 | 3646 | XP_006720856.1 | |
AF105366.1 | 20 | 2911 | AAD39742.1 | |
AF108831.1 | 20 | 2645 | AAD25337.1 | |
AF116242.1 | 20 | 2747 | AAF24986.1 | |
AF477977.1 | 21 | 3104 | AAL85335.1 | |
AF531258.1 | 19 | 2866 | AAQ10026.1 | |
AF531259.1 | 21 | 3104 | AAQ10027.1 | |
AF531260.1 | 21 | 2742 | AAQ10028.1 | |
AK128133.1 | 19 | 2419 | ||
AK292550.1 | 21 | 2850 | ||
AK315283.1 | 20 | 2708 | ||
AL117500.1 | 17 | 2339 | CAB55965.1 | |
BC051744.1 | 21 | 3100 | AAH51744.1 | |
BC070107.1 | 21 | 2767 | ||
BC126241.1 | 20 | 2644 | ||
BC126243.1 | 20 | 2644 | ||
BX648195.1 | 20 | 3783 | CAI46042.1 | |
DQ138323.1 | 21 | 3162 | ABA02873.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1052620 | Chr.15:32612402 - 34538731 on Build GRCh38 | Gain | EMC4 GOLGA8A AVEN TMCO5B LOC105370757 MIR1233-2 GREM1 ARHGAP11A NUTM1 EMC7 LOC101928134 SCG5 LPCAT4 LOC100131315 FMN1 CHRM5 MIR1233-1 PGBD4 GOLGA8B KATNBL1 RYR3 LOC101928042 NOP10 SLC12A6 |
esv33337 | Chr.15:31689863 - 34575097 on Build GRCh38 | Gain+Loss | WHAMMP1 AVEN LOC105370757 MIR1233-2 GREM1 CHRNA7 LOC100653133 ULK4P1 LOC100996255 NUTM1 EMC7 LOC101928134 FMN1 GOLGA8O LOC105370940 PGBD4 GOLGA8B RYR3 LOC101928042 GOLGA8N SLC12A6 GOLGA8K EMC4 GOLGA8A TMCO5B LOC101060588 ARHGAP11A SCG5 LPCAT4 LOC100131315 CHRM5 MIR1233-1 KATNBL1 NOP10 |
esv3569431 | Chr.15:33564570 - 35080307 on Build GRCh38 | Gain | EMC4 GOLGA8A AVEN MIR1233-2 NUTM1 GJD2 ACTC1 EMC7 AQR LOC101928174 LPCAT4 CHRM5 MIR1233-1 ZNF770 PGBD4 GOLGA8B KATNBL1 RYR3 NOP10 SLC12A6 |
dgv4456n54 | Chr.15:32604149 - 34557061 on Build GRCh38 | Gain | AVEN LOC105370757 MIR1233-2 GREM1 NUTM1 EMC7 LOC101928134 FMN1 PGBD4 GOLGA8B RYR3 LOC101928042 GOLGA8N SLC12A6 EMC4 GOLGA8A TMCO5B ARHGAP11A SCG5 LPCAT4 LOC100131315 CHRM5 MIR1233-1 KATNBL1 NOP10 |
Set Membership: |
Intragenic Exonic DGV Variation |