Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 4125 |
Gene Name: | mannosidase alpha class 2B member 1 |
Gene Aliases: |
LAMAN, MANB |
Location: |
Chr.19:12646508-12666777 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAN2B1 | NM_000528.3 | NP_000519.2 | ||
NM_001173498.1 | NP_001166969.1 | |||
XM_005259913.1 | XP_005259970.1 | |||
XM_017026818.1 | XP_016882307.1 | |||
AB209921.1 | BAD93158.1 | |||
AK291572.1 | ||||
AK303418.1 | ||||
AW172353.1 | ||||
BC000736.2 | AAH00736.1 | |||
BM982008.1 | ||||
U05572.1 | AAB03816.1 | |||
U60266.1 | AAC34130.1 | |||
U68382.1 | AAC50811.1 | |||
U68567.1 | AAC50812.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160586 | Chr.19:12597480 - 12823909 on Build GRCh38 | Duplication | HOOK2 ASNA1 SNORD135 ZNF791 JUNB TNPO2 FBXW9 BEST2 C19orf43 DHPS MIR5684 ZNF490 LOC105372280 WDR83OS SNORD41 PRDX2 WDR83 RNASEH2A MAN2B1 |
dgv138n111 | Chr.19:12627853 - 12810372 on Build GRCh38 | Deletion | HOOK2 ASNA1 SNORD135 ZNF791 JUNB TNPO2 FBXW9 BEST2 C19orf43 DHPS MIR5684 LOC105372280 WDR83OS SNORD41 PRDX2 WDR83 RNASEH2A MAN2B1 |
Set Membership: |
Intragenic Non-exonic DGV Variation |