Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 284058 |
Gene Name: | KAT8 regulatory NSL complex subunit 1 |
Gene Aliases: |
CENP-36, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1 |
Location: |
Chr.17:46029916-46225374 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KANSL1 | NM_001193465.1 | 16 | 3552 | NP_001180394.1 |
NM_001193466.1 | 15 | 3769 | NP_001180395.1 | |
NM_015443.3 | 15 | 3471 | NP_056258.1 | |
XM_006721823.1 | 16 | 3555 | XP_006721886.1 | |
XM_006721824.3 | 16 | 3739 | XP_006721887.1 | |
XM_011524628.2 | 16 | 3736 | XP_011522930.1 | |
XM_011524629.2 | 15 | 3637 | XP_011522931.1 | |
XM_011524630.2 | 15 | 3550 | XP_011522932.1 | |
XM_011524631.2 | 15 | 3548 | XP_011522933.1 | |
XM_011524632.2 | 15 | 2165 | XP_011522934.1 | |
XM_017024488.1 | XP_016879977.1 | |||
AB033093.1 | 14 | 3392 | BAA86581.1 | |
AK094946.1 | 7 | 1270 | ||
AK291259.1 | 15 | 3529 | ||
AK310569.1 | 1 | 1071 | ||
AL117476.1 | 4 | 639 | CAB55949.1 | |
AL137317.1 | 11 | 1713 | CAB70694.1 | |
BC035892.1 | 10 | 1576 | AAH35892.1 | |
BC098376.1 | 15 | 3471 | AAH98376.1 | |
BX538006.1 | 15 | 3772 | CAD97958.1 | |
BX648760.1 | 8 | 7529 | CAH10565.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471698 | Chr.17:46002113 - 46300887 on Build GRCh38 | Gain+Loss | MAPT LRRC37A KANSL1-AS1 ARL17B KANSL1 |
nsv469705 | Chr.17:45969304 - 46127121 on Build GRCh38 | Loss | MAPT STH KANSL1 |
nsv469537 | Chr.17:46002112 - 46163353 on Build GRCh38 | Gain+Loss | MAPT KANSL1 |
nsv575142 | Chr.17:46003698 - 46172742 on Build GRCh38 | Loss | MAPT KANSL1 |
esv2762433 | Chr.17:45496768 - 46710828 on Build GRCh38 | Gain+Loss | MAPT NSFP1 LOC644172 LOC107987253 LRRC37A4P SPPL2C STH MAPT-AS1 KANSL1-AS1 KANSL1 NSF LOC105376839 CRHR1 LRRC37A MGC57346-CRHR1 MAPT-IT1 ARL17A MGC57346 LOC107987254 ARL17B CRHR1-IT1 LRRC37A2 |
nsv1072740 | Chr.17:45970433 - 46248034 on Build GRCh38 | Deletion | MAPT STH KANSL1-AS1 KANSL1 |
dgv3178n100 | Chr.17:45877004 - 46034461 on Build GRCh38 | Gain | MAPT MAPT-IT1 STH MAPT-AS1 KANSL1 |
esv2751686 | Chr.17:45962198 - 46286690 on Build GRCh38 | Gain | MAPT LRRC37A STH KANSL1-AS1 ARL17B KANSL1 |
esv2758692 | Chr.17:46006548 - 47199967 on Build GRCh38 | Gain+Loss | MAPT RPS2P47 MIR5089 NSFP1 LOC107987253 KANSL1-AS1 KANSL1 NSF LRRC37A WNT9B WNT3 ARL17A RPRML GOSR2 LOC107987254 ARL17B LRRC37A2 CDC27 |
dgv128e55 | Chr.17:46017105 - 46286690 on Build GRCh38 | Gain | MAPT LRRC37A KANSL1-AS1 ARL17B KANSL1 |
dgv127e55 | Chr.17:45927606 - 46733750 on Build GRCh38 | Gain | MAPT LRRC37A NSFP1 LOC107987253 ARL17A STH KANSL1-AS1 LOC107987254 ARL17B LRRC37A2 KANSL1 NSF |
Set Membership: |
Intragenic Exonic DGV Variation |