Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3082 |
Gene Name: | hepatocyte growth factor |
Gene Aliases: |
DFNB39, F-TCF, HGFB, HPTA, SF |
Location: |
Chr.7:81699006-81770438 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HGF | NM_000601.5 | NP_000592.3 | ||
NM_001010931.2 | 8 | 1240 | NP_001010931.1 | |
NM_001010932.2 | NP_001010932.1 | |||
NM_001010933.2 | 8 | 1225 | NP_001010933.1 | |
XM_006715956.2 | XP_006716019.1 | |||
XM_011516115.2 | XP_011514417.1 | |||
XM_017012097.1 | 9 | 1203 | XP_016867586.1 | |
XM_017012098.1 | 9 | 1188 | XP_016867587.1 | |
AK291674.1 | ||||
AK313694.1 | ||||
BC022308.1 | 8 | 1074 | AAH22308.1 | |
BC130284.1 | ||||
BC130286.1 | ||||
BM971901.1 | 1 | 130 | ||
L02931.1 | 8 | 1073 | AAA52649.1 | |
M29145.1 | AAA52650.1 | |||
M55379.1 | ||||
M60718.1 | AAA52648.1 | |||
M73239.1 | AAA64239.1 | |||
M73240.1 | AAA64297.1 | |||
M77227.1 | 8 | 1096 | AAA35980.1 | |
X16323.1 | CAA34387.1 | |||
X57574.1 | 8 | 1090 | CAA40802.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1019753 | Chr.7:81692852 - 82052256 on Build GRCh38 | Gain | LOC101927356 HGF CACNA2D1 |
Set Membership: |
Intragenic Exonic DGV Variation |