Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 8398 |
Gene Name: | phospholipase A2 group VI |
Gene Aliases: |
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta |
Location: |
Chr.22:38111495-38192099 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G6 | NM_001004426.1 | 16 | 2497 | NP_001004426.1 |
NM_001199562.1 | 16 | 2452 | NP_001186491.1 | |
NM_003560.2 | 17 | 2659 | NP_003551.2 | |
XM_005261764.2 | 17 | 2634 | XP_005261821.1 | |
XM_005261765.1 | 17 | 2631 | XP_005261822.1 | |
XM_005261766.1 | 17 | 2726 | XP_005261823.1 | |
XM_006724332.3 | 17 | 2635 | XP_006724395.1 | |
XM_011530422.1 | 17 | 2510 | XP_011528724.1 | |
XM_011530423.1 | 18 | 2746 | XP_011528725.1 | |
XM_011530424.1 | 19 | 2870 | XP_011528726.1 | |
XM_011530425.1 | 17 | 2570 | XP_011528727.1 | |
XM_017028981.1 | 16 | 2564 | XP_016884470.1 | |
XM_017028982.1 | 16 | 2469 | XP_016884471.1 | |
XM_017028983.1 | 18 | 2708 | XP_016884472.1 | |
XM_017028984.1 | 17 | 2584 | XP_016884473.1 | |
XM_017028985.1 | 16 | 2402 | XP_016884474.1 | |
AF064594.1 | 17 | 2659 | AAC97486.1 | |
AF102988.1 | 15 | 2361 | AAD41722.1 | |
AF102989.1 | 16 | 2523 | AAD41723.1 | |
AK096521.1 | 16 | 2192 | ||
AK128862.1 | 6 | 3415 | ||
AK290825.1 | 16 | 2454 | ||
AK291212.1 | 17 | 2645 | ||
AL080187.1 | 17 | 2659 | CAB45768.2 | |
BC036742.1 | 17 | 2653 | AAH36742.2 | |
BC051904.1 | 16 | 2433 | AAH51904.1 | |
CR456543.1 | CAG30429.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471198 | Chr.22:38019714 - 38149612 on Build GRCh38 | Loss | PLA2G6 POLR2F BAIAP2L2 SLC16A8 PICK1 |
esv996111 | Chr.22:38108982 - 38112324 on Build GRCh38 | Insertion | PLA2G6 BAIAP2L2 |
nsv3623 | Chr.22:38105535 - 38113801 on Build GRCh38 | Insertion | PLA2G6 BAIAP2L2 |
nsv508732 | Chr.22:38096355 - 38180191 on Build GRCh38 | Insertion | PLA2G6 BAIAP2L2 |
nsv834194 | Chr.22:38063495 - 38240504 on Build GRCh38 | Loss | PLA2G6 TMEM184B BAIAP2L2 SLC16A8 SNORA92 MAFF PICK1 |
nsv834193 | Chr.22:37997329 - 38172090 on Build GRCh38 | Loss | PLA2G6 POLR2F BAIAP2L2 SLC16A8 PICK1 |
Set Membership: |
Intragenic Exonic DGV Variation |