Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 100526739 |
Gene Name: | APITD1-CORT readthrough |
Gene Aliases: |
- |
Location: |
Chr.1:10430102-10452003 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APITD1-CORT | NM_001243768.1 | NP_001230697.1 | ||
NM_001270517.1 | NP_001257446.1 | |||
NM_198544.3 | NP_940946.1 | |||
NM_199006.2 | NP_950171.2 | |||
NR_037187.1 | ||||
AA858169.1 | ||||
AF516753.1 | AAM61954.1 | |||
AF521016.1 | AAM70482.1 | |||
CF128105.1 | ||||
HY054077.1 |
Entrez Gene ID: | 1325 |
Gene Name: | cortistatin |
Gene Aliases: |
CST-14, CST-17, CST-29 |
Location: |
Chr.1:10449719-10452003 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CORT | NM_001302.4 | 1 | 224 | NP_001293.3 |
AF086433.1 | ||||
AY358561.1 | 1 | 224 | AAQ89950.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2764192 | Chr.1:10323676 - 10534129 on Build GRCh38 | Gain | DFFA KIF1B CORT PEX14 APITD1 APITD1-CORT PGD |
esv2758920 | Chr.1:10178012 - 10522497 on Build GRCh38 | Gain+Loss | DFFA KIF1B CORT PEX14 APITD1 APITD1-CORT UBE4B PGD MIR1273D |
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |