Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 9776 |
Gene Name: | autophagy related 13 |
Gene Aliases: |
KIAA0652, PARATARG8 |
Location: |
Chr.11:46617276-46676019 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 23 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3626191 | Chr.11:46651662 - 46681651 on Build GRCh38 | Gain | ATG13 ARHGAP1 |
Set Membership: |
Intragenic Non-exonic DGV Variation |