Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3172 |
Gene Name: | hepatocyte nuclear factor 4 alpha |
Gene Aliases: |
FRTS4, HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14 |
Location: |
Chr.20:44355801-44432845 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HNF4A | NM_000457.4 | 2 | 361 | NP_000448.3 |
NM_001030003.2 | 2 | 182 | NP_001025174.1 | |
NM_001030004.2 | 2 | 182 | NP_001025175.1 | |
NM_001258355.1 | 3 | 452 | NP_001245284.1 | |
NM_001287182.1 | 3 | 404 | NP_001274111.1 | |
NM_001287183.1 | 3 | 404 | NP_001274112.1 | |
NM_001287184.1 | 3 | 404 | NP_001274113.1 | |
NM_175914.4 | 2 | 182 | NP_787110.2 | |
NM_178849.2 | 2 | 361 | NP_849180.1 | |
NM_178850.2 | 2 | 361 | NP_849181.1 | |
XM_005260407.3 | 3 | 481 | XP_005260464.1 | |
AB307703.1 | 2 | 217 | ||
AY680696.1 | 2 | 182 | AAT91237.1 | |
AY680697.1 | 2 | 182 | AAT91238.1 | |
AY680698.1 | 2 | 182 | AAT91239.1 | |
BC137539.1 | 2 | 283 | ||
BC137540.1 | 2 | 283 | ||
BP274413.1 | 2 | 361 | ||
FJ608822.1 | 3 | 400 | ||
HQ692860.1 | 2 | 244 | ||
HQ692868.1 | 2 | 178 | ||
HQ692869.1 | 2 | 244 | ||
X76930.1 | 2 | 236 | CAA54248.1 | |
X87870.1 | 2 | 316 | CAA61133.1 | |
X87871.1 | 2 | 301 | CAA61134.1 | |
X87872.1 | 2 | 337 | CAA61135.1 | |
Z49825.1 | 3 | 437 | CAA89989.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1057847 | Chr.20:42652464 - 44431352 on Build GRCh38 | Loss | PTPRT OSER1 LOC101927159 L3MBTL1 R3HDML SGK2 TOX2 LOC101927200 OSER1-AS1 JPH2 MYBL2 IFT52 MIR3646 SRSF6 LOC105372626 GDAP1L1 HNF4A-AS1 FITM2 HNF4A LOC101927138 GTSF1L |
nsv586065 | Chr.20:44261867 - 44505244 on Build GRCh38 | Gain | TTPAL LINC01430 MIR3646 R3HDML GDAP1L1 HNF4A-AS1 FITM2 LINC01620 SERINC3 HNF4A |
Set Membership: |
Intragenic Exonic DGV Variation |