Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 23230 |
Gene Name: | vacuolar protein sorting 13 homolog A |
Gene Aliases: |
CHAC, CHOREIN |
Location: |
Chr.9:77177353-77421537 on Build GRCh38 |
Assay Gene Location: | Within Exon 74 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VPS13A | NM_001018037.1 | 71 | 10636 | NP_001018047.1 |
NM_033305.2 | 72 | 10753 | NP_150648.2 | |
AF337532.1 | 72 | 10753 | AAK61861.1 | |
AJ608769.1 | 71 | 10636 | CAE75581.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1051079 | Chr.9:77361673 - 77471809 on Build GRCh38 | Gain | GNA14-AS1 VPS13A GNA14 |
nsv1041647 | Chr.9:77211923 - 77537136 on Build GRCh38 | Gain | GNA14-AS1 VPS13A GNA14 |
esv3620773 | Chr.9:77386177 - 77418202 on Build GRCh38 | Loss | VPS13A |
Set Membership: |
Intragenic Exonic DGV Variation |