Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2153 |
Gene Name: | coagulation factor V |
Gene Aliases: |
FVL, PCCF, RPRGL1, THPH2 |
Location: |
Chr.1:169511954-169586632 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
F5 | NM_000130.4 | 13 | 3127 | NP_000121.2 |
XM_017000660.1 | 12 | 3136 | XP_016856149.1 | |
AJ297255.1 | 1 | 1007 | CAC82573.1 | |
AK226079.1 | 13 | 3045 | ||
AK291613.1 | 13 | 3077 | ||
M14335.1 | 13 | 3058 | AAB59532.1 | |
M16967.1 | 13 | 3072 | AAA52424.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv22143 | Chr.1:169090390 - 169652030 on Build GRCh38 | Gain+Loss | CCDC181 ATP1B1 SLC19A2 LOC101928596 BLZF1 F5 SELP NME7 |
Set Membership: |
Intragenic Exonic DGV Variation |