Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 145173 |
Gene Name: | beta 3-glucosyltransferase |
Gene Aliases: |
B3GALTL, B3GTL, B3Glc-T, Gal-T, beta3Glc-T |
Location: |
Chr.13:31199975-31332276 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
B3GLCT | NM_194318.3 | 15 | 2264 | NP_919299.3 |
XM_006719768.3 | 16 | 2454 | XP_006719831.1 | |
XM_011534936.1 | 14 | 2133 | XP_011533238.1 | |
XM_011534938.2 | 14 | 2102 | XP_011533240.1 | |
XM_017020395.1 | 16 | 2357 | XP_016875884.1 | |
AK094979.1 | 2 | 910 | ||
AK291273.1 | 15 | 2253 | ||
AY190526.1 | 15 | 2154 | AAO37647.1 | |
BC068595.1 | 15 | 2244 | AAH68595.1 | |
BX647932.1 | 2 | 1330 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2751142 | Chr.13:31152041 - 31332004 on Build GRCh38 | Gain | HSPH1 B3GLCT |
nsv561400 | Chr.13:31297526 - 31682595 on Build GRCh38 | Gain | B3GLCT LOC105370149 |
Set Membership: |
Intragenic Exonic DGV Variation |