Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2189 |
Gene Name: | Fanconi anemia complementation group G |
Gene Aliases: |
FAG, XRCC9 |
Location: |
Chr.9:35073838-35080016 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FANCG | NM_004629.1 | NP_004620.1 | ||
AJ007669.1 | CAA07602.1 | |||
AK293427.1 | ||||
AK311348.1 | ||||
AK312987.1 | ||||
BC000032.2 | AAH00032.1 | |||
BC011623.2 | AAH11623.1 | |||
BT009813.1 | AAP88815.1 | |||
KU159422.1 | ||||
KU178053.1 | ||||
KU178054.1 | ||||
KU178055.1 | ||||
KU178056.1 | ||||
KU178057.1 | ||||
U70310.1 | AAB80802.1 |
Set Membership: |
Intragenic Non-exonic non-DGV Variation |