Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 55135 |
Gene Name: | WD repeat containing antisense to TP53 |
Gene Aliases: |
DKCB3, TCAB1, WDR79 |
Location: |
Chr.17:7686071-7703502 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WRAP53 | NM_001143990.1 | NP_001137462.1 | ||
NM_001143991.1 | NP_001137463.1 | |||
NM_001143992.1 | 1 | 139 | NP_001137464.1 | |
NM_018081.2 | 1 | 139 | NP_060551.2 | |
XM_017024812.1 | 1 | 916 | XP_016880301.1 | |
AK001247.1 | BAA91579.1 | |||
AK056669.1 | 1 | 1808 | ||
AK092370.1 | 1 | 888 | ||
AK225273.1 | ||||
AK225411.1 | ||||
AK303246.1 | ||||
AY766322.1 | ||||
AY766323.1 | 1 | 139 | ||
BC002336.2 | AAH02336.1 | |||
DQ431240.1 | 1 | 139 | ||
DQ431241.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss | YBX2 ZBTB4 FXR2 MIR497HG SNORA67 TP53 ASGR1 RNASEK BCL6B SLC35G6 TNFSF12 ACADVL CLDN7 TNFSF12-TNFSF13 MIR324 CHRNB1 ALOX12 SAT2 SLC16A13 GPS2 KCTD11 POLR2A PHF23 MIR497 TMEM256-PLSCR3 MPDU1 LOC100996842 NEURL4 CD68 ATP1B2 C17orf49 CTDNEP1 SNORA48 WRAP53 ALOX12-AS1 GABARAP EIF5A ELP5 LOC107983988 MIR195 TMEM102 TMEM256 SOX15 C17orf74 SENP3-EIF4A1 SNORD10 SHBG TNK1 ASGR2 SLC2A4 TMEM95 DLG4 RNASEK-C17orf49 FGF11 SENP3 SLC16A11 ACAP1 EIF4A1 SPEM1 TNFSF13 CLEC10A PLSCR3 NLGN2 DVL2 |
Set Membership: |
Intragenic Exonic DGV Variation |