Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 89781 |
Gene Name: | HPS4, biogenesis of lysosomal organelles complex 3 subunit 2 |
Gene Aliases: |
BLOC3S2, LE |
Location: |
Chr.22:26443613-26483863 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Set Membership: |
Intragenic Exonic non-DGV Variation |