Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 23500 |
Gene Name: | dishevelled associated activator of morphogenesis 2 |
Gene Aliases: |
dJ90A20A.1 |
Location: |
Chr.6:39792366-39904877 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DAAM2 | NM_001201427.1 | 25 | 5646 | NP_001188356.1 |
NM_015345.3 | 25 | 5707 | NP_056160.2 | |
XM_006715039.3 | 26 | 6318 | XP_006715102.3 | |
XM_006715040.3 | 25 | 6290 | XP_006715103.3 | |
XM_006715042.2 | 26 | 8313 | XP_006715105.1 | |
XM_006715043.2 | 26 | 5690 | XP_006715106.1 | |
XM_006715045.3 | 26 | 6905 | XP_006715108.1 | |
XM_006715046.3 | 26 | 6083 | XP_006715109.1 | |
XM_017010630.1 | 26 | 6929 | XP_016866119.1 | |
AB002379.2 | 25 | 5646 | ||
AL833083.2 | 25 | 5687 | CAD89973.1 | |
BX647839.1 | 9 | 6229 |
Entrez Gene ID: | 4337 |
Gene Name: | molybdenum cofactor synthesis 1 |
Gene Aliases: |
MIG11, MOCOD, MOCODA |
Location: |
Chr.6:39904258-39934551 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MOCS1 | NM_001075098.3 | 11 | 4080 | NP_001068566.1 |
NM_005943.5 | 10 | 4084 | NP_005934.2 | |
NR_033233.1 | 9 | 3890 | ||
XM_011514633.2 | 10 | 3928 | XP_011512935.1 | |
XM_017010888.1 | 10 | 3898 | XP_016866377.1 | |
AV724791.1 | 1 | 80 | ||
BC036839.1 | AAH36839.1 | |||
CA944911.1 | 1 | 86 |
Set Membership: |
Intragenic Exonic non-DGV Variation |