Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 23230 |
Gene Name: | vacuolar protein sorting 13 homolog A |
Gene Aliases: |
CHAC, CHOREIN |
Location: |
Chr.9:77177353-77421537 on Build GRCh38 |
Assay Gene Location: | Within Exon 39 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VPS13A | NM_001018037.1 | 38 | 4928 | NP_001018047.1 |
NM_001018038.2 | 39 | 5045 | NP_001018048.1 | |
NM_015186.3 | 39 | 5045 | NP_056001.1 | |
NM_033305.2 | 39 | 5045 | NP_150648.2 | |
AB054005.1 | 39 | 4956 | BAB59128.1 | |
AF337532.1 | 39 | 5045 | AAK61861.1 | |
AJ608769.1 | 38 | 4928 | CAE75581.1 | |
AJ626859.1 | 39 | 5045 | CAF25186.1 | |
AK000273.1 | 9 | 1374 | ||
AK124024.1 | 14 | 2075 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv614729 | Chr.9:77291835 - 77366058 on Build GRCh38 | Loss | VPS13A |
nsv1041647 | Chr.9:77211923 - 77537136 on Build GRCh38 | Gain | GNA14-AS1 GNA14 VPS13A |
nsv614728 | Chr.9:77195973 - 77375037 on Build GRCh38 | Loss | VPS13A |
Set Membership: |
Intragenic Exonic DGV Variation |