Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 23098 |
Gene Name: | sterile alpha and TIR motif containing 1 |
Gene Aliases: |
MyD88-5, SAMD2, SARM |
Location: |
Chr.17:28371662-28401045 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SARM1 | NM_015077.3 | 9 | 6088 | NP_055892.2 |
AB011096.1 | 9 | 5340 | BAA25450.1 | |
AJ290445.1 | 9 | 5643 | CAB90355.1 | |
BG820141.1 | 1 | 441 |
Entrez Gene ID: | 113235 |
Gene Name: | solute carrier family 46 member 1 |
Gene Aliases: |
G21, HCP1, PCFT |
Location: |
Chr.17:28394642-28406630 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC46A1 | NM_001242366.2 | NP_001229295.1 | ||
NM_080669.5 | NP_542400.2 | |||
XM_005277786.3 | XP_005277843.1 | |||
XM_017024110.1 | XP_016879599.1 | |||
AK054669.1 | BAB70789.1 | |||
AK074161.1 | BAB84987.1 | |||
AK097194.1 | ||||
AL832613.2 | CAD89945.1 | |||
BC010691.1 | AAH10691.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1062056 | Chr.17:28376191 - 28452023 on Build GRCh38 | Gain | SLC46A1 SARM1 |
nsv1056839 | Chr.17:28348199 - 28428188 on Build GRCh38 | Gain | TMEM199 MIR4723 VTN SLC46A1 POLDIP2 SEBOX SARM1 |
nsv574653 | Chr.17:28227749 - 28501479 on Build GRCh38 | Gain | KRT18P55 MIR4723 VTN LOC105371712 PPY2P TMEM199 PYY2 LOC105371930 RPS7P1 SLC46A1 TNFAIP1 IFT20 POLDIP2 SLC13A2 LOC101926984 SEBOX SARM1 TMEM97 |
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |