Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 90678 |
Gene Name: | leucine rich repeat and sterile alpha motif containing 1 |
Gene Aliases: |
CMT2P, RIFLE, TAL |
Location: |
Chr.9:127451486-127503501 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRSAM1 | NM_001005373.3 | NP_001005373.1 | ||
NM_001190723.2 | NP_001177652.1 | |||
NM_138361.5 | 1 | 408 | NP_612370.3 | |
XM_006717316.3 | XP_006717379.1 | |||
XM_017015283.1 | XP_016870772.1 | |||
XM_017015284.1 | XP_016870773.1 | |||
AK056203.1 | BAB71119.1 | |||
AK091589.1 | 1 | 156 | BAC03703.1 | |
AK226106.1 | ||||
AY358830.1 | AAQ89189.1 | |||
DA165729.1 | ||||
DA393757.1 | 1 | 384 | ||
DA726792.1 | 1 | 412 | ||
DB165274.1 | ||||
Z44095.1 | 1 | 143 |
Set Membership: |
Intragenic Exonic non-DGV Variation |