Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 23400 |
Gene Name: | ATPase 13A2 |
Gene Aliases: |
CLN12, HSA9947, KRPPD, PARK9 |
Location: |
Chr.1:16985958-17011972 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 29 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508947 | Chr.1:16909817 - 17011115 on Build GRCh38 | Insertion | CROCC MFAP2 ATP13A2 |
nsv428421 | Chr.1:16437838 - 17157486 on Build GRCh38 | Gain+Loss | ESPNP CROCC LOC105376796 LOC105376805 NECAP2 LOC102723383 FAM231B RNU1-1 MFAP2 RNU1-4 PADI2 SPATA21 MST1L CROCCP2 RNU1-3 RNU1-2 SDHB NBPF1 MIR3675 CROCCP3 ATP13A2 FAM231A MST1P2 LOC100132147 |
nsv954854 | Chr.1:16961106 - 17013905 on Build GRCh38 | Deletion | CROCC MFAP2 ATP13A2 |
dgv108n100 | Chr.1:16850538 - 17010370 on Build GRCh38 | Gain | CROCC LOC105376805 RNU1-2 MIR3675 MFAP2 ATP13A2 |
dgv92n100 | Chr.1:16684943 - 17003642 on Build GRCh38 | Gain | ESPNP CROCC LOC105376805 MST1L RNU1-2 LOC102723383 MIR3675 MFAP2 ATP13A2 RNU1-4 |
nsv834280 | Chr.1:16888218 - 17123006 on Build GRCh38 | Loss | CROCC PADI2 LOC105376805 RNU1-2 SDHB MFAP2 ATP13A2 |
dgv120n100 | Chr.1:16890896 - 16997938 on Build GRCh38 | Gain | CROCC RNU1-2 MFAP2 ATP13A2 |
nsv834269 | Chr.1:16855283 - 17038531 on Build GRCh38 | Loss | CROCC LOC105376805 RNU1-2 SDHB MIR3675 MFAP2 ATP13A2 |
Set Membership: |
Intragenic Non-exonic DGV Variation |