Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 55063 |
Gene Name: | zinc finger CW-type and PWWP domain containing 1 |
Gene Aliases: |
ZCW1 |
Location: |
Chr.7:100400826-100429239 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZCWPW1 | NM_001258008.1 | 4 | 550 | NP_001244937.1 |
NM_017984.4 | 4 | 550 | NP_060454.3 | |
XM_005250479.2 | 4 | 981 | XP_005250536.1 | |
XM_005250480.2 | 4 | 981 | XP_005250537.1 | |
XM_006716035.3 | 4 | 981 | XP_006716098.1 | |
XM_006716036.2 | 4 | 485 | XP_006716099.1 | |
XM_006716037.3 | 4 | 981 | XP_006716100.1 | |
XM_006716038.3 | 4 | 981 | XP_006716101.1 | |
XM_006716040.3 | 4 | 981 | XP_006716103.1 | |
XM_017012379.1 | 4 | 981 | XP_016867868.1 | |
XM_017012380.1 | 4 | 485 | XP_016867869.1 | |
AK093038.1 | 4 | 485 | BAC04028.1 | |
AK299989.1 | 4 | 438 | ||
AK300745.1 | 4 | 406 | ||
AK302108.1 | 4 | 404 | ||
AK316553.1 | 4 | 738 | ||
AL136735.1 | 4 | 421 | CAB66669.1 | |
BC002725.1 | 1 | 139 | AAH02725.1 | |
DC413605.1 | 4 | 465 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | PMS2P1 PPP1R35 TRIM56 VGF CNPY4 MUC17 MIR6840 MIR106B MUC12 ACTL6B AP4M1 AZGP1 GAL3ST4 MUC3A NAT16 PCOLCE-AS1 PLOD3 ZAN TRIP6 ZASP MEPCE ACHE PILRA TAF6 ZNHIT1 MCM7 STAG3L5P GNB2 ZSCAN21 SRRT FIS1 PILRB MIR6875 TSC22D4 LAMTOR4 ZNF3 SAP25 STAG3 COPS6 EPHB4 STAG3L5P-PVRIG2P-PILRB NYAP1 EPO SPDYE3 PVRIG MIR93 MBLAC1 ZKSCAN1 MIR25 MIR4653 SLC12A9 GIGYF1 MIR4658 ZCWPW1 LOC105375429 MOSPD3 PCOLCE UFSP1 C7orf61 AGFG2 AP1S1 POP7 LRCH4 PVRIG2P TFR2 MOGAT3 GPC2 C7orf43 LOC102724094 FBXO24 GATS CLDN15 AZGP1P1 SERPINE1 |
nsv831071 | Chr.7:100320402 - 100515790 on Build GRCh38 | Loss | PMS2P1 PPP1R35 MEPCE STAG3L5P-PVRIG2P-PILRB C7orf61 PILRA NYAP1 STAG3L5P PVRIG2P SPDYE3 MIR6840 PILRB TSC22D4 ZCWPW1 |
nsv527107 | Chr.7:100417088 - 100467096 on Build GRCh38 | Loss | PPP1R35 MEPCE C7orf61 TSC22D4 ZCWPW1 |
Set Membership: |
Intragenic Exonic DGV Variation |