Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 50943 |
Gene Name: | forkhead box P3 |
Gene Aliases: |
AIID, DIETER, IPEX, JM2, PIDX, XPID |
Location: |
Chr.X:49250436-49266505 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FOXP3 | NM_001114377.1 | NP_001107849.1 | ||
NM_014009.3 | NP_054728.2 | |||
XM_006724533.2 | XP_006724596.2 | |||
XM_011543916.2 | XP_011542218.1 | |||
XM_017029565.1 | XP_016885054.1 | |||
XM_017029566.1 | XP_016885055.1 | |||
XM_017029567.1 | XP_016885056.1 | |||
AF277993.1 | AAG53607.1 | |||
AJ005891.1 | CAA06748.1 | |||
AK292052.1 | ||||
AK299988.1 | ||||
BC113401.1 | ||||
BC113403.1 | ||||
BC143785.1 | ||||
BC143786.1 | ||||
DQ010327.1 | AAY27088.1 | |||
EF534714.1 | ||||
EU855812.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471602 | Chr.X:49205142 - 49320261 on Build GRCh38 | Gain | CCDC22 PPP1R3F FOXP3 CACNA1F GAGE10 |
nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain | WAS WDR45 SYP-AS1 CCDC120 LOC101927635 PIM2 PRICKLE3 KCND1 RBM3 GPKOW CCDC22 SUV39H1 FOXP3 HDAC6 TFE3 PORCN WDR13 PLP2 SLC38A5 OTUD5 GRIPAP1 FTSJ1 PCSK1N TIMM17B CACNA1F PRAF2 MAGIX EBP GLOD5 TBC1D25 GATA1 PPP1R3F PQBP1 SLC35A2 ERAS SYP |
esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss | SSX6 WAS WDR45 SSX4B SYP-AS1 ZNF182 CCDC120 LOC101927635 PIM2 SSX1 PRICKLE3 ZNF630-AS1 KCND1 RBM3 GPKOW SSX4 CCDC22 SUV39H1 FOXP3 HDAC6 TFE3 SSX3 PORCN WDR13 PLP2 SLC38A5 OTUD5 GRIPAP1 SPACA5B FTSJ1 PCSK1N TIMM17B CACNA1F PRAF2 SPACA5 SSX5 MAGIX EBP SSX9 GLOD5 TBC1D25 GATA1 PPP1R3F PQBP1 SLC35A2 ZNF630 ERAS SYP |
nsv469734 | Chr.X:49205141 - 49320261 on Build GRCh38 | Gain+Loss | CCDC22 PPP1R3F FOXP3 CACNA1F GAGE10 |
nsv6895 | Chr.X:49227319 - 49262690 on Build GRCh38 | Insertion | CCDC22 FOXP3 CACNA1F |
Set Membership: |
Intragenic Non-exonic DGV Variation |