Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 7133 |
Gene Name: | TNF receptor superfamily member 1B |
Gene Aliases: |
CD120b, TBPII, TNF-R-II, TNF-R75, TNFBR, TNFR1B, TNFR2, TNFR80, p75, p75TNFR |
Location: |
Chr.1:12166948-12209222 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TNFRSF1B | NM_001066.2 | 10 | 2859 | NP_001057.1 |
XM_011542060.2 | 11 | 2983 | XP_011540362.1 | |
XM_011542063.2 | 10 | 2917 | XP_011540365.1 | |
XM_017002214.1 | 10 | 2883 | XP_016857703.1 | |
XM_017002215.1 | 9 | 2814 | XP_016857704.1 | |
BC052977.1 | 10 | 2857 | AAH52977.1 | |
M32315.1 | 10 | 2859 | AAA59929.1 | |
S63368.1 | 9 | 2662 | AAB19824.2 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471622 | Chr.1:12155207 - 12371588 on Build GRCh38 | Gain+Loss | TNFRSF1B VPS13D MIR4632 MIR7846 |
nsv508891 | Chr.1:12097920 - 12235045 on Build GRCh38 | Insertion | TNFRSF1B TNFRSF8 VPS13D MIR4632 MIR7846 |
esv3584475 | Chr.1:12134394 - 12217607 on Build GRCh38 | Gain | TNFRSF1B TNFRSF8 MIR4632 MIR7846 |
Set Membership: |
Intragenic Exonic DGV Variation |