Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 90678 |
Gene Name: | leucine rich repeat and sterile alpha motif containing 1 |
Gene Aliases: |
CMT2P, RIFLE, TAL |
Location: |
Chr.9:127451486-127503501 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 21 - Intron 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRSAM1 | NM_001005373.3 | NP_001005373.1 | ||
NM_001005374.3 | NP_001005374.1 | |||
NM_001190723.2 | NP_001177652.1 | |||
NM_138361.5 | NP_612370.3 | |||
XM_006717316.3 | XP_006717379.1 | |||
XM_017015283.1 | XP_016870772.1 | |||
XM_017015284.1 | XP_016870773.1 | |||
AK056203.1 | BAB71119.1 | |||
AK056305.1 | BAB71144.1 | |||
AK091589.1 | BAC03703.1 | |||
AK226106.1 | ||||
AY358830.1 | AAQ89189.1 | |||
BC009239.2 | AAH09239.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv466569 | Chr.9:127470144 - 127512011 on Build GRCh38 | Loss | FAM129B LRSAM1 |
Set Membership: |
Intragenic Non-exonic DGV Variation |