Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2904 |
Gene Name: | glutamate ionotropic receptor NMDA type subunit 2B |
Gene Aliases: |
EIEE27, GluN2B, MRD6, NMDAR2B, NR2B, hNR3 |
Location: |
Chr.12:13537337-13982012 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRIN2B | XM_005253351.3 | 4 | 3750 | XP_005253408.1 |
XM_011520628.2 | 14 | 6816 | XP_011518930.1 | |
XM_011520629.2 | 15 | 6623 | XP_011518931.1 | |
XM_017019219.1 | 14 | 6371 | XP_016874708.1 | |
U88963.1 | 13 | 6023 | AAD00659.1 |
Set Membership: |
Intragenic Exonic non-DGV Variation |