Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2072 |
Gene Name: | ERCC excision repair 4, endonuclease catalytic subunit |
Gene Aliases: |
ERCC11, FANCQ, RAD1, XFEPS, XPF |
Location: |
Chr.16:13920138-13952348 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ERCC4 | NM_005236.2 | NP_005227.1 | ||
XM_011522424.2 | XP_011520726.1 | |||
XM_011522427.1 | XP_011520729.1 | |||
XM_017023043.1 | XP_016878532.1 | |||
AK289726.1 | ||||
AK301930.1 | ||||
U64315.1 | AAB07689.1 |
Set Membership: |
Intragenic Non-exonic non-DGV Variation |