Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2153 |
Gene Name: | coagulation factor V |
Gene Aliases: |
FVL, PCCF, RPRGL1, THPH2 |
Location: |
Chr.1:169511954-169586632 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
F5 | NM_000130.4 | NP_000121.2 | ||
XM_017000660.1 | XP_016856149.1 | |||
AK226079.1 | ||||
AK291613.1 | ||||
AK300469.1 | ||||
M14335.1 | AAB59532.1 | |||
M16967.1 | AAA52424.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv22143 | Chr.1:169090390 - 169652030 on Build GRCh38 | Gain+Loss | LOC101928596 NME7 SLC19A2 F5 BLZF1 CCDC181 SELP ATP1B1 |
esv2518089 | Chr.1:169555099 - 169556054 on Build GRCh38 | Insertion | F5 |
Set Membership: |
Intragenic Non-exonic DGV Variation |