Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 4089 |
Gene Name: | SMAD family member 4 |
Gene Aliases: |
DPC4, JIP, MADH4, MYHRS |
Location: |
Chr.18:51030213-51085042 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMAD4 | NM_005359.5 | NP_005350.1 | ||
AK290770.1 | ||||
BC002379.2 | AAH02379.1 | |||
BX647129.1 | ||||
KU178168.1 | ||||
KU178169.1 | ||||
U44378.1 | AAA91041.1 |
Set Membership: |
Intragenic Non-exonic non-DGV Variation |