Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 5351 |
Gene Name: | procollagen-lysine,2-oxoglutarate 5-dioxygenase 1 |
Gene Aliases: |
EDS6, LH, LH1, LLH, PLOD |
Location: |
Chr.1:11934667-11975542 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLOD1 | NM_000302.3 | 19 | 2674 | NP_000293.2 |
NM_001316320.1 | 20 | 2815 | NP_001303249.1 | |
AI347587.1 | 1 | 358 | ||
AK124035.1 | 2 | 1988 | ||
BC016657.2 | 19 | 2578 | AAH16657.1 | |
BM991136.1 | 1 | 374 | ||
BQ447373.1 | 1 | 374 | ||
M98252.1 | 19 | 2585 |
Set Membership: |
Intragenic Exonic non-DGV Variation |