Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 7157 |
Gene Name: | tumor protein p53 |
Gene Aliases: |
BCC7, LFS1, P53, TRP53 |
Location: |
Chr.17:7668402-7687550 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TP53 | NM_000546.5 | 1 | 43 | NP_000537.3 |
NM_001126112.2 | 1 | 43 | NP_001119584.1 | |
NM_001126113.2 | 1 | 43 | NP_001119585.1 | |
NM_001126114.2 | 1 | 43 | NP_001119586.1 | |
NM_001126118.1 | 1 | 43 | NP_001119590.1 | |
NM_001276695.1 | 1 | 43 | NP_001263624.1 | |
NM_001276696.1 | 1 | 43 | NP_001263625.1 | |
NM_001276760.1 | 1 | 43 | NP_001263689.1 | |
NM_001276761.1 | 1 | 43 | NP_001263690.1 | |
AF307851.1 | AAG28785.1 | |||
AK225838.1 | ||||
AK297462.1 | ||||
AK297927.1 | ||||
AK303277.1 | ||||
AK312568.1 | ||||
AY627884.1 | AAT40418.1 | |||
DA308036.1 | ||||
DA453049.1 | 1 | 43 | ||
DA932053.1 | 1 | 38 | ||
DC347346.1 | ||||
DQ186648.1 | ABA29753.1 | |||
DQ186649.1 | ABA29754.1 | |||
DQ191317.1 | ABB80262.1 | |||
DQ286964.1 | ABB80266.1 | |||
FJ207420.1 | ||||
JN214348.1 | ||||
JN900492.1 | ||||
M14694.1 | AAA61211.1 | |||
X02469.1 | CAA26306.1 |
Entrez Gene ID: | 55135 |
Gene Name: | WD repeat containing antisense to TP53 |
Gene Aliases: |
DKCB3, TCAB1, WDR79 |
Location: |
Chr.17:7686071-7703502 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WRAP53 | NM_001143990.1 | NP_001137462.1 | ||
NM_001143991.1 | 1 | 69 | NP_001137463.1 | |
AK056669.1 | 1 | 829 | ||
AY766322.1 | 1 | 69 | ||
DQ431241.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss | TNK1 C17orf74 SLC16A13 NLGN2 GPS2 SNORA48 DLG4 ACADVL LOC100996842 GABARAP LOC107983988 SLC16A11 CLDN7 SHBG TNFSF12 TP53 MPDU1 SENP3-EIF4A1 TMEM102 ALOX12 SNORA67 CTDNEP1 DVL2 MIR497 C17orf49 FGF11 SLC2A4 RNASEK MIR497HG RNASEK-C17orf49 KCTD11 ATP1B2 TMEM95 EIF5A CHRNB1 ASGR1 TNFSF13 SENP3 YBX2 ASGR2 BCL6B SOX15 CD68 ALOX12-AS1 MIR324 SPEM1 SLC35G6 MIR195 PLSCR3 TMEM256-PLSCR3 ACAP1 EIF4A1 WRAP53 TMEM256 SNORD10 NEURL4 FXR2 ELP5 POLR2A PHF23 TNFSF12-TNFSF13 SAT2 ZBTB4 CLEC10A |
Set Membership: |
Intragenic Exonic DGV Variation |