Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 146057 |
Gene Name: | tau tubulin kinase 2 |
Gene Aliases: |
SCA11, TTBK |
Location: |
Chr.15:42738974-42920995 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TTBK2 | NM_173500.3 | 14 | 2739 | NP_775771.3 |
XM_005254171.4 | 13 | 2873 | XP_005254228.1 | |
XM_005254173.4 | 13 | 2621 | XP_005254230.1 | |
XM_006720402.3 | 13 | 2348 | XP_006720465.1 | |
XM_006720403.3 | 14 | 2515 | XP_006720466.1 | |
XM_017021950.1 | 12 | 2364 | XP_016877439.1 | |
AB020654.1 | 2 | 504 | BAA74870.1 | |
AF525400.1 | 17 | 3918 | AAO14996.1 | |
AK131372.1 | 14 | 2463 | BAD18523.1 | |
BC071556.1 | 15 | 2513 | AAH71556.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528725 | Chr.15:42463357 - 42812208 on Build GRCh38 | Gain | LRRC57 SNAP23 STARD9 HAUS2 ZNF106 CDAN1 TTBK2 |
nsv457122 | Chr.15:42656664 - 42770350 on Build GRCh38 | Loss | STARD9 CDAN1 TTBK2 |
Set Membership: |
Intragenic Exonic DGV Variation |