Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 79147 |
Gene Name: | fukutin related protein |
Gene Aliases: |
LGMD2I, MDC1C, MDDGA5, MDDGB5, MDDGC5 |
Location: |
Chr.19:46746015-46758575 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FKRP | NM_001039885.2 | 4 | 1183 | NP_001034974.1 |
NM_024301.4 | 4 | 1131 | NP_077277.1 | |
XM_005259247.2 | 5 | 1244 | XP_005259304.1 | |
XM_005259248.2 | 5 | 1234 | XP_005259305.1 | |
XM_005259249.4 | 5 | 1176 | XP_005259306.1 | |
XM_011527306.1 | 3 | 980 | XP_011525608.1 | |
XM_011527307.1 | 4 | 1029 | XP_011525609.1 | |
XM_017027297.1 | 4 | 1350 | XP_016882786.1 | |
AJ314847.1 | 4 | 1119 | CAC85633.1 | |
AK022638.1 | 4 | 1119 | BAB14146.1 | |
AK095497.1 | 4 | 1170 | ||
AK291282.1 | 4 | 1119 | ||
BC002612.2 | 2 | 927 | AAH02612.1 | |
BM979509.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833850 | Chr.19:46703285 - 46867053 on Build GRCh38 | Loss | FKRP LOC107987269 SNAR-E PRKD2 STRN4 SLC1A5 MIR320E ARHGAP35 AP2S1 |
Set Membership: |
Intragenic Exonic DGV Variation |