Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 4684 |
Gene Name: | neural cell adhesion molecule 1 |
Gene Aliases: |
CD56, MSK39, NCAM |
Location: |
Chr.11:112961247-113278436 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NCAM1 | NM_000615.6 | 19 | 4472 | NP_000606.3 |
NM_001242607.1 | 21 | 4580 | NP_001229536.1 | |
NM_181351.4 | 20 | 4502 | NP_851996.2 | |
AL832563.1 | 2 | 2118 | ||
BC014205.2 | 14 | 3366 | AAH14205.2 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv483080 | Chr.11:113216072 - 113400474 on Build GRCh38 | Loss | NCAM1-AS1 TTC12 NCAM1 ANKK1 |
Set Membership: |
Intragenic Exonic DGV Variation |