Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 4625 |
Gene Name: | myosin, heavy chain 7, cardiac muscle, beta |
Gene Aliases: |
CMD1S, CMH1, MPD1, MYHCB, SPMD, SPMM |
Location: |
Chr.14:23412738-23435686 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 38 - Exon 38 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYH7 | NM_000257.3 | NP_000248.2 | ||
XM_017021340.1 | XP_016876829.1 | |||
AB209708.1 | BAD92945.1 | |||
AJ710171.1 | ||||
AY518538.1 | AAT44534.1 | |||
BC112171.1 | AAI12172.1 | |||
BC112173.1 | AAI12174.1 | |||
DQ248310.1 | ABB55265.1 | |||
EF560725.1 | ||||
EU747717.1 | ||||
M17712.1 | AAA36343.1 | |||
M21665.1 | AAA36345.1 | |||
M58018.1 | AAA62830.1 | |||
X03741.1 | CAA27381.1 | |||
X05631.1 | CAA29119.1 | |||
X06976.1 | CAA30039.1 | |||
X51591.1 | CAA35940.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470623 | Chr.14:23391499 - 23418656 on Build GRCh38 | Gain | MIR208B MHRT MYH7 MYH6 |
esv2748480 | Chr.14:23390535 - 23420438 on Build GRCh38 | Deletion | MIR208B MHRT MYH7 MYH6 |
nsv456166 | Chr.14:23392602 - 23418656 on Build GRCh38 | Gain | MIR208B MHRT MYH7 MYH6 |
esv3584149 | Chr.14:23395407 - 23427942 on Build GRCh38 | Gain | MIR208B MHRT MYH7 MYH6 |
Set Membership: |
Intragenic Non-exonic DGV Variation |