Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 4594 |
Gene Name: | methylmalonyl-CoA mutase |
Gene Aliases: |
MCM |
Location: |
Chr.6:49430360-49463328 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MUT | NM_000255.3 | NP_000246.2 | ||
XM_005249143.3 | XP_005249200.1 | |||
AK292568.1 | ||||
AK312611.1 | ||||
BC016282.1 | AAH16282.1 | |||
BT007434.1 | AAP36102.1 | |||
BX647789.1 | ||||
KF030882.1 | ||||
KU178201.1 | ||||
KU178202.1 | ||||
M65131.1 | AAA59569.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5960n100 | Chr.6:49322472 - 49684411 on Build GRCh38 | Gain | CRISP2 MUT C6orf141 RHAG GLYATL3 CENPQ |
dgv10654n54 | Chr.6:49453528 - 49478796 on Build GRCh38 | Gain | MUT CENPQ |
Set Membership: |
Intragenic Non-exonic DGV Variation |