Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 401237 |
Gene Name: | cancer susceptibility candidate 15 (non-protein coding) |
Gene Aliases: |
LINC00340 |
Location: |
Chr.6:21666444-22194400 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CASC15 | NR_015410.1 | |||
AK022865.1 | ||||
AK026189.1 | ||||
KP981381.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830602 | Chr.6:21973618 - 22165178 on Build GRCh38 | Gain | NBAT1 CASC15 |
nsv1161343 | Chr.6:21861465 - 22037804 on Build GRCh38 | Deletion | CASC15 |
esv3570802 | Chr.6:21959225 - 22038891 on Build GRCh38 | Loss | CASC15 |