Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2645 |
Gene Name: | glucokinase |
Gene Aliases: |
FGQTL3, GK, GLK, HHF3, HK4, HKIV, HXKP, LGLK, MODY2 |
Location: |
Chr.7:44143213-44189439 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GCK | NM_000162.3 | NP_000153.1 | ||
XM_017011966.1 | XP_016867455.1 | |||
BC001890.1 | AAH01890.1 | |||
BT007062.1 | AAP35711.1 | |||
EU008605.1 | ||||
M88011.1 | AAA51824.1 | |||
M90299.1 | AAA52562.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830983 | Chr.7:44068842 - 44201624 on Build GRCh38 | Loss | YKT6 POLD2 MIR6838 GCK MIR4649 AEBP1 MYL7 POLM |
nsv8082 | Chr.7:43956156 - 44207229 on Build GRCh38 | Gain | SPDYE1 POLR2J4 LINC00957 RASA4CP AEBP1 MYL7 MIR6837 YKT6 POLD2 MIR6838 GCK PGAM2 MIR4649 DBNL POLM |
nsv509195 | Chr.7:44147118 - 44261782 on Build GRCh38 | Insertion | YKT6 GCK CAMK2B |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |