Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 54714 |
Gene Name: | cyclic nucleotide gated channel beta 3 |
Gene Aliases: |
ACHM1 |
Location: |
Chr.8:86573608-86743675 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CNGB3 | NM_019098.4 | NP_061971.3 | ||
XM_011517138.2 | XP_011515440.1 | |||
AF228520.1 | AAF80179.1 | |||
AF272900.1 | AAF86274.1 | |||
BC150601.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422239 | Chr.8:86535165 - 87814698 on Build GRCh38 | Duplication | CPNE3 CNBD1 CNGB3 |
nsv1023510 | Chr.8:86416772 - 86862720 on Build GRCh38 | Loss | CPNE3 RMDN1 WWP1 CNGB3 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |