Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 10658 |
Gene Name: | CUGBP, Elav-like family member 1 |
Gene Aliases: |
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50 |
Location: |
Chr.11:47465937-47565569 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CELF1 | NM_006560.3 | NP_006551.1 | ||
XM_011519855.1 | XP_011518157.1 | |||
XM_011519858.1 | XP_011518160.1 | |||
XM_017017115.1 | XP_016872604.1 | |||
XM_017017116.1 | XP_016872605.1 | |||
XM_017017118.1 | XP_016872607.1 | |||
XM_017017119.1 | XP_016872608.1 | |||
XM_017017121.1 | XP_016872610.1 | |||
XM_017017123.1 | XP_016872612.1 | |||
XM_017017127.1 | XP_016872616.1 | |||
XM_017017128.1 | XP_016872617.1 | |||
XM_017017131.1 | XP_016872620.1 | |||
XM_017017132.1 | XP_016872621.1 | |||
CB960956.1 | ||||
DA320861.1 | ||||
DB162286.1 | ||||
U63289.1 | AAC50895.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832142 | Chr.11:47496403 - 47659758 on Build GRCh38 | Loss | PTPMT1 NDUFS3 MTCH2 C1QTNF4 AGBL2 CELF1 KBTBD4 FAM180B |
nsv314 | Chr.11:47540012 - 47584704 on Build GRCh38 | Deletion | PTPMT1 NDUFS3 CELF1 KBTBD4 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |