Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 10658 |
Gene Name: | CUGBP, Elav-like family member 1 |
Gene Aliases: |
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50 |
Location: |
Chr.11:47465937-47565569 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832142 | Chr.11:47496403 - 47659758 on Build GRCh38 | Loss | KBTBD4 CELF1 NDUFS3 AGBL2 C1QTNF4 FAM180B MTCH2 PTPMT1 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |