Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 8398 |
Gene Name: | phospholipase A2 group VI |
Gene Aliases: |
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta |
Location: |
Chr.22:38111495-38192099 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471198 | Chr.22:38019714 - 38149612 on Build GRCh38 | Loss | BAIAP2L2 POLR2F SLC16A8 PICK1 PLA2G6 |
nsv1059531 | Chr.22:38124868 - 38169202 on Build GRCh38 | Loss | PLA2G6 |
nsv508732 | Chr.22:38096355 - 38180191 on Build GRCh38 | Insertion | BAIAP2L2 PLA2G6 |
nsv834194 | Chr.22:38063495 - 38240504 on Build GRCh38 | Loss | SNORA92 BAIAP2L2 SLC16A8 TMEM184B PICK1 MAFF PLA2G6 |
nsv834193 | Chr.22:37997329 - 38172090 on Build GRCh38 | Loss | BAIAP2L2 POLR2F SLC16A8 PICK1 PLA2G6 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |