Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2187 |
Gene Name: | Fanconi anemia complementation group B |
Gene Aliases: |
FA2, FAAP90, FAAP95, FAB, FACB |
Location: |
Chr.X:14690863-14873255 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FANCB | NM_001018113.2 | NP_001018123.1 | ||
NM_001324162.1 | NP_001311091.1 | |||
NM_152633.3 | NP_689846.1 | |||
XM_011545470.2 | XP_011543772.1 | |||
XM_017029355.1 | XP_016884844.1 | |||
XM_017029356.1 | XP_016884845.1 | |||
AK091383.1 | BAC03650.1 | |||
BC043596.1 | AAH43596.1 | |||
BC136558.1 | ||||
BC136560.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6812 | Chr.X:14836320 - 14868456 on Build GRCh38 | Deletion | FANCB |
nsv9934 | Chr.X:14730738 - 15173654 on Build GRCh38 | Loss | MOSPD2 GLRA2 FANCB |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |