Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 101929680 |
Gene Name: | uncharacterized LOC101929680 |
Gene Aliases: |
- |
Location: |
Chr.2:166081531-166301784 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101929680 | NR_110260.1 | |||
KM096550.1 | ||||
KM096551.1 |
Entrez Gene ID: | 6335 |
Gene Name: | sodium voltage-gated channel alpha subunit 9 |
Gene Aliases: |
ETHA, FEB3B, GEFSP7, HSAN2D, NE-NA, NENA, Nav1.7, PN1, SFNP |
Location: |
Chr.2:166195185-166375987 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SCN9A | NM_002977.3 | NP_002968.1 | ||
XM_005246757.2 | XP_005246814.1 | |||
XM_011511616.2 | XP_011509918.1 | |||
XM_011511617.2 | XP_011509919.1 | |||
XM_011511618.2 | XP_011509920.1 | |||
XM_011511619.2 | XP_011509921.1 | |||
XM_017004668.1 | XP_016860157.1 | |||
XM_017004669.1 | XP_016860158.1 | |||
DQ857292.1 | ||||
X82835.1 | CAA58042.1 |
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |