Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 25953 |
Gene Name: | paroxysmal nonkinesigenic dyskinesia |
Gene Aliases: |
BRP17, DYT8, FKSG19, FPD1, KIPP1184, MR-1, MR1, PDC, PKND1, TAHCCP2 |
Location: |
Chr.2:218270392-218346793 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PNKD | NM_001077399.2 | NP_001070867.1 | ||
NM_015488.4 | NP_056303.3 | |||
XM_017003771.1 | XP_016859260.1 | |||
AB033010.1 | BAA86498.1 | |||
AF318057.1 | AAL25716.1 | |||
AF417001.1 | AAL08573.1 | |||
AF418285.1 | AAL08580.1 | |||
AK289867.1 | ||||
AL080092.1 | CAB45707.2 | |||
BC021118.1 | AAH21118.1 | |||
BC036457.2 | AAH36457.1 | |||
BQ028777.1 |
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |