Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 6622 |
Gene Name: | synuclein alpha |
Gene Aliases: |
NACP, PARK1, PARK4, PD1 |
Location: |
Chr.4:89724099-89838324 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1012406 | Chr.4:89246630 - 90245636 on Build GRCh38 | Loss | SNCA MMRN1 CCSER1 SNCA-AS1 GPRIN3 |
nsv594806 | Chr.4:88621130 - 90539537 on Build GRCh38 | Gain | FAM13A SNCA TIGD2 HERC3 MMRN1 CCSER1 FAM13A-AS1 SNCA-AS1 GPRIN3 NAP1L5 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |