Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3082 |
Gene Name: | hepatocyte growth factor |
Gene Aliases: |
DFNB39, F-TCF, HGFB, HPTA, SF |
Location: |
Chr.7:81699006-81770438 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv464609 | Chr.7:81748600 - 81781600 on Build GRCh38 | Loss | HGF |
nsv1019753 | Chr.7:81692852 - 82052256 on Build GRCh38 | Gain | HGF CACNA2D1 LOC101927356 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |