Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 55063 |
Gene Name: | zinc finger CW-type and PWWP domain containing 1 |
Gene Aliases: |
ZCW1 |
Location: |
Chr.7:100400826-100429239 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZCWPW1 | NM_001258008.1 | NP_001244937.1 | ||
NM_017984.4 | NP_060454.3 | |||
XM_005250479.2 | XP_005250536.1 | |||
XM_005250480.2 | XP_005250537.1 | |||
XM_006716035.3 | XP_006716098.1 | |||
XM_006716036.2 | XP_006716099.1 | |||
XM_006716037.3 | XP_006716100.1 | |||
XM_006716038.3 | XP_006716101.1 | |||
XM_006716040.3 | XP_006716103.1 | |||
XM_017012379.1 | XP_016867868.1 | |||
XM_017012380.1 | XP_016867869.1 | |||
AK000919.1 | BAA91424.1 | |||
AK093038.1 | BAC04028.1 | |||
AK300745.1 | ||||
AK302108.1 | ||||
AL136735.1 | CAB66669.1 | |||
BC002725.1 | AAH02725.1 | |||
KU178677.1 | ||||
KU178678.1 | ||||
KU178679.1 | ||||
KU178680.1 | ||||
KU178681.1 | ||||
KU178682.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | GNB2 ZCWPW1 MIR4658 ZASP LRCH4 ACTL6B MIR106B AP1S1 CLDN15 VGF NYAP1 MOGAT3 MUC12 MIR25 GPC2 ZAN AZGP1P1 SLC12A9 PVRIG2P PMS2P1 PCOLCE EPHB4 TRIM56 STAG3L5P-PVRIG2P-PILRB C7orf43 UFSP1 PCOLCE-AS1 ZKSCAN1 ZNF3 MUC17 TSC22D4 TRIP6 MUC3A ZNHIT1 SERPINE1 TAF6 NAT16 SPDYE3 MIR93 CNPY4 MOSPD3 MIR4653 SRRT POP7 STAG3L5P PVRIG PILRA AZGP1 PPP1R35 SAP25 EPO MBLAC1 STAG3 GIGYF1 PILRB LOC105375429 FIS1 COPS6 GATS AP4M1 MIR6875 MEPCE PLOD3 MCM7 MIR6840 ACHE AGFG2 LOC102724094 FBXO24 C7orf61 ZSCAN21 TFR2 LAMTOR4 GAL3ST4 |
nsv831071 | Chr.7:100320402 - 100515790 on Build GRCh38 | Loss | PVRIG2P ZCWPW1 PMS2P1 PILRB STAG3L5P-PVRIG2P-PILRB SPDYE3 MEPCE NYAP1 MIR6840 STAG3L5P PILRA TSC22D4 C7orf61 PPP1R35 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |