Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 1956 |
Gene Name: | epidermal growth factor receptor |
Gene Aliases: |
ERBB, ERBB1, HER1, NISBD2, PIG61, mENA |
Location: |
Chr.7:55019032-55207338 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EGFR | NM_005228.3 | NP_005219.2 | ||
AB209442.1 | BAD92679.1 | |||
AK225422.1 | ||||
AK290352.1 | ||||
AK294750.1 | ||||
AU137334.1 | ||||
AY550966.1 | AAT52212.1 | |||
BC094761.1 | AAH94761.1 | |||
DQ088980.1 | ||||
GU255993.1 | ||||
HQ912715.1 | ||||
K03193.1 | AAA52371.1 | |||
X00588.1 | CAA25240.1 | |||
X00663.1 | CAA25282.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734425 | Chr.7:53868335 - 55917254 on Build GRCh38 | Deletion | LOC101060341 EGFR FKBP9P1 LANCL2 VSTM2A-OT1 PSPHP1 VSTM2A ZNF713 HPVC1 LINC01445 LOC100996654 EGFR-AS1 ELDR LOC107986797 SEC61G VOPP1 SEPT14 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |