Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3082 |
Gene Name: | hepatocyte growth factor |
Gene Aliases: |
DFNB39, F-TCF, HGFB, HPTA, SF |
Location: |
Chr.7:81699006-81770438 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HGF | NM_000601.5 | NP_000592.3 | ||
NM_001010932.2 | NP_001010932.1 | |||
XM_006715956.2 | XP_006716019.1 | |||
XM_011516115.2 | XP_011514417.1 | |||
AK291674.1 | ||||
AK313694.1 | ||||
BC130284.1 | ||||
BC130286.1 | ||||
M29145.1 | AAA52650.1 | |||
M55379.1 | ||||
M60718.1 | AAA52648.1 | |||
M73239.1 | AAA64239.1 | |||
M73240.1 | AAA64297.1 | |||
X16323.1 | CAA34387.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3576398 | Chr.7:81685626 - 81732874 on Build GRCh38 | Gain | LOC100128317 HGF |
nsv1019753 | Chr.7:81692852 - 82052256 on Build GRCh38 | Gain | LOC101927356 CACNA2D1 HGF |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |