Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 1956 |
Gene Name: | epidermal growth factor receptor |
Gene Aliases: |
ERBB, ERBB1, HER1, NISBD2, PIG61, mENA |
Location: |
Chr.7:55019032-55207338 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EGFR | NM_005228.3 | NP_005219.2 | ||
NM_201282.1 | NP_958439.1 | |||
NM_201283.1 | NP_958440.1 | |||
NM_201284.1 | NP_958441.1 | |||
AB209442.1 | BAD92679.1 | |||
AF125253.1 | AAG43240.1 | |||
AK225422.1 | ||||
AK290352.1 | ||||
AW163038.1 | ||||
AY550966.1 | AAT52212.1 | |||
AY698023.1 | AAT97978.1 | |||
AY698024.1 | AAT97979.1 | |||
BC094761.1 | AAH94761.1 | |||
DQ088980.1 | ||||
GU255993.1 | ||||
HQ912715.1 | ||||
K03193.1 | AAA52371.1 | |||
U48722.1 | AAC50802.1 | |||
U95089.1 | AAB53063.1 | |||
X00588.1 | CAA25240.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2761332 | Chr.7:54574291 - 55097556 on Build GRCh38 | Gain | LOC107986797 LOC100996654 SEC61G EGFR |
nsv607029 | Chr.7:54579095 - 55086688 on Build GRCh38 | Gain | LOC107986797 LOC100996654 SEC61G EGFR |
nsv5753 | Chr.7:55045323 - 55070844 on Build GRCh38 | Insertion | EGFR |
esv2734425 | Chr.7:53868335 - 55917254 on Build GRCh38 | Deletion | LINC01445 VSTM2A-OT1 LOC100996654 FKBP9P1 SEC61G ZNF713 LOC101060341 VOPP1 EGFR-AS1 EGFR VSTM2A LOC107986797 SEPT14 ELDR LANCL2 PSPHP1 HPVC1 |
nsv1030676 | Chr.7:54583600 - 55151489 on Build GRCh38 | Gain | LOC107986797 LOC100996654 SEC61G EGFR |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |